Print…
Anti-Sodium Potassium ATPase Mouse Monoclonal Antibody [clone: EP1845Y]
Fornecedor: Abcam
AB283318-100 AB283318-20UL AB283318-1ML
ABCAAB283318-100EA 475 EUR
ABCAAB283318-100 ABCAAB283318-20UL ABCAAB283318-1ML
Anti-Sodium Potassium ATPase Mouse Monoclonal Antibody [clone: EP1845Y]
Anticorpos
Mouse monoclonal [EP1845Y] to Sodium Potassium ATPase - Mouse IgG1.
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Sodium Potassium ATPase
Clonality: Monoclonal
Clone: EP1845Y
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human; Mouse; Rat