Print…

Anti-Heme Oxygenase 1 Mouse Monoclonal Antibody [clone: 23-Heme Oxygenase 1]

Fornecedor: Abcam
AB302672-1 AB302672-100
ABCAAB302672-1EA 9160 EUR
ABCAAB302672-1 ABCAAB302672-100
Anti-Heme Oxygenase 1 Mouse Monoclonal Antibody [clone: 23-Heme Oxygenase 1]
Anticorpos
Mouse monoclonal [23/Heme Oxygenase 1] to Heme Oxygenase 1 - BSA and Azide free.

Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Heme Oxygenase 1
Clonality: Monoclonal
Clone: 23-Heme Oxygenase 1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR