Print…

Anti-GRO gamma+GCP2 Rabbit Monoclonal Antibody [clone: EPR22310-120]

Fornecedor: Abcam
AB259523-100 AB259523-1
ABCAAB259523-100EA 869 EUR
ABCAAB259523-100 ABCAAB259523-1
Anti-GRO gamma+GCP2 Rabbit Monoclonal Antibody [clone: EPR22310-120]
Anticorpos
Rabbit monoclonal [EPR22310-120] to GRO gamma + GCP2 - BSA and Azide free (Detector).

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: GRO gamma+GCP2
Clonality: Monoclonal
Clone: EPR22310-120
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR