Print…

Anti-eNOS Rabbit Polyclonal Antibody

Fornecedor: Abcam
AB230455-100 AB230455-1
ABCAAB230455-100EA 663 EUR
ABCAAB230455-100 ABCAAB230455-1
Anti-eNOS Rabbit Polyclonal Antibody
Anticorpos
Rabbit polyclonal to eNOS - C-terminal.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: eNOS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR