Print…

Anti-Decorin Rabbit Monoclonal Antibody [clone: EPR24097-105]

Fornecedor: Abcam
AB277636-100 AB277636-20UL AB277636-1ML
ABCAAB277636-100EA 663 EUR
ABCAAB277636-100 ABCAAB277636-20UL ABCAAB277636-1ML
Anti-Decorin Rabbit Monoclonal Antibody [clone: EPR24097-105]
Anticorpos
Rabbit monoclonal [EPR24097-105] to Decorin.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Decorin
Clonality: Monoclonal
Clone: EPR24097-105
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR