Print…
Anti-MDC1 Rabbit Monoclonal Antibody [clone: EPR24360-116]
Fornecedor: Abcam
AB271061-100 AB271061-20UL AB271061-1ML
ABCAAB271061-100EA 713 EUR
ABCAAB271061-100 ABCAAB271061-20UL ABCAAB271061-1ML
Anti-MDC1 Rabbit Monoclonal Antibody [clone: EPR24360-116]
Anticorpos
Rabbit monoclonal [EPR24360-116] to MDC1.
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: MDC1
Clonality: Monoclonal
Clone: EPR24360-116
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human