Print…

Anti-Rab3A Rabbit Monoclonal Antibody [clone: EPR24450-44]

Fornecedor: Abcam
AB285163-100 AB285163-20UL AB285163-1ML
ABCAAB285163-100EA 713 EUR
ABCAAB285163-100 ABCAAB285163-20UL ABCAAB285163-1ML
Anti-Rab3A Rabbit Monoclonal Antibody [clone: EPR24450-44]
Anticorpos
Rabbit monoclonal [EPR24450-44] to Rab3A.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Rab3A
Clonality: Monoclonal
Clone: EPR24450-44
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR