Print…

Anti-Monoamine Oxidase A/MAO-A Rabbit Monoclonal Antibody [clone: EPR7101]

Fornecedor: Abcam
AB240031-100UG AB240031-1
ABCAAB240031-100UGEA 1020 EUR
ABCAAB240031-100UG ABCAAB240031-1
Anti-Monoamine Oxidase A/MAO-A Rabbit Monoclonal Antibody [clone: EPR7101]
Anticorpos
Rabbit monoclonal [EPR7101] to Monoamine Oxidase A/MAO-A - BSA and Azide free.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Monoamine Oxidase A/MAO-A
Clonality: Monoclonal
Clone: EPR7101
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat; Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR