Print…

Anti-Rho guanine exchange factor 16/NBR Rabbit Monoclonal Antibody [clone: EPR9950]

Fornecedor: Abcam
AB249097-100 AB249097-1
ABCAAB249097-100EA 1020 EUR
ABCAAB249097-100 ABCAAB249097-1
Anti-Rho guanine exchange factor 16/NBR Rabbit Monoclonal Antibody [clone: EPR9950]
Anticorpos
Rabbit monoclonal [EPR9950] to Rho guanine exchange factor 16/NBR - BSA and Azide free.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Rho guanine exchange factor 16/NBR
Clonality: Monoclonal
Clone: EPR9950
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR