Print…

Anti-Angiotensin Converting Enzyme 1 Rabbit Monoclonal Antibody [clone: EPR22291-247]

Fornecedor: Abcam
AB254222-100 AB254222-20UL AB254222-1ML
ABCAAB254222-100EA 813 EUR
ABCAAB254222-100 ABCAAB254222-20UL ABCAAB254222-1ML
Anti-Angiotensin Converting Enzyme 1 Rabbit Monoclonal Antibody [clone: EPR22291-247]
Anticorpos
Rabbit monoclonal [EPR22291-247] to Angiotensin Converting Enzyme 1.

Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Angiotensin Converting Enzyme 1
Clonality: Monoclonal
Clone: EPR22291-247
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR