Print…
Anti-Niemann Pick C2 Rabbit Monoclonal Antibody [clone: EPR19993-145-1]
Fornecedor: Abcam
AB251560-100 AB251560-1
ABCAAB251560-100EA 1020 EUR
ABCAAB251560-100 ABCAAB251560-1
Anti-Niemann Pick C2 Rabbit Monoclonal Antibody [clone: EPR19993-145-1]
Anticorpos
Rabbit monoclonal [EPR19993-145-1] to Niemann Pick C2 - BSA and Azide free.
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Niemann Pick C2
Clonality: Monoclonal
Clone: EPR19993-145-1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat