Print…

Anti-Vwf Rabbit Monoclonal Antibody [clone: EPR25069-131]

Fornecedor: Abcam
AB287967-1 AB287967-100
ABCAAB287967-1EA 9160 EUR
ABCAAB287967-1 ABCAAB287967-100
Anti-Vwf Rabbit Monoclonal Antibody [clone: EPR25069-131]
Anticorpos
Rabbit monoclonal [EPR25069-131] to Von Willebrand Factor - BSA and Azide free.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Vwf
Clonality: Monoclonal
Clone: EPR25069-131
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR