Print…

Anti-ZNFN1A2/HELIOS Rabbit Monoclonal Antibody [clone: EPR23200-40]

Fornecedor: Abcam
AB252411-100 AB252411-20UL AB252411-1ML
ABCAAB252411-100EA 675 EUR
ABCAAB252411-100 ABCAAB252411-20UL ABCAAB252411-1ML
Anti-ZNFN1A2/HELIOS Rabbit Monoclonal Antibody [clone: EPR23200-40]
Anticorpos
Rabbit monoclonal [EPR23200-40] to ZNFN1A2/HELIOS.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ZNFN1A2/HELIOS
Clonality: Monoclonal
Clone: EPR23200-40
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR