Print…

Anti-Quinone oxidoreductase Rabbit Monoclonal Antibody [clone: EPR10843(B)]

Fornecedor: Abcam
AB154842-100 AB154842-1ML
ABCAAB154842-100EA 744 EUR
ABCAAB154842-100 ABCAAB154842-1ML
Anti-Quinone oxidoreductase Rabbit Monoclonal Antibody [clone: EPR10843(B)]
Anticorpos
Rabbit monoclonal [EPR10843(B)] to Quinone oxidoreductase.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Quinone oxidoreductase
Clonality: Monoclonal
Clone: EPR10843(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR