Print…
Anti-Vitamin B2 Rabbit Monoclonal Antibody [clone: EPR20853-141]
Fornecedor: Abcam
AB254211-100 AB254211-1
ABCAAB254211-100EA 1020 EUR
ABCAAB254211-100 ABCAAB254211-1
Anti-Vitamin B2 Rabbit Monoclonal Antibody [clone: EPR20853-141]
Anticorpos
Rabbit monoclonal [EPR20853-141] to Vitamin B2 - BSA and Azide free.
Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Vitamin B2
Clonality: Monoclonal
Clone: EPR20853-141
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar