Print…

Anti-Cellubrevin Rabbit Monoclonal Antibody [clone: EPR25065-16]

Fornecedor: Abcam
AB283312-100 AB283312-20UL AB283312-1ML
ABCAAB283312-100EA 713 EUR
ABCAAB283312-100 ABCAAB283312-20UL ABCAAB283312-1ML
Anti-Cellubrevin Rabbit Monoclonal Antibody [clone: EPR25065-16]
Anticorpos
Rabbit monoclonal [EPR25065-16] to Cellubrevin.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cellubrevin
Clonality: Monoclonal
Clone: EPR25065-16
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR