Print…

Anti-Sodium Potassium ATPase Rat Monoclonal Antibody [clone: EP1845Y]

Fornecedor: Abcam
AB283345-100 AB283345-20UL AB283345-1ML
ABCAAB283345-100EA 475 EUR
ABCAAB283345-100 ABCAAB283345-20UL ABCAAB283345-1ML
Anti-Sodium Potassium ATPase Rat Monoclonal Antibody [clone: EP1845Y]
Anticorpos
Rat monoclonal [EP1845Y] to Sodium Potassium ATPase - Rat IgG2a.

Type: Primary
Antigen: Sodium Potassium ATPase
Clonality: Monoclonal
Clone: EP1845Y
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human; Mouse; Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR