Print…
Anti-Tau (phospho T217) Rabbit Monoclonal Antibody [clone: EPR24654-19]
Fornecedor: Abcam
AB314478-100UL AB314478-20UL AB314478-1ML
ABCAAB314478-100ULEA 694 EUR
ABCAAB314478-100UL ABCAAB314478-20UL ABCAAB314478-1ML
Anti-Tau (phospho T217) Rabbit Monoclonal Antibody [clone: EPR24654-19]
Anticorpos
Rabbit monoclonal [EPR24654-19] to Tau (phospho T217).
Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Tau
Clonality: Monoclonal
Clone: EPR24654-19
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar