Print…

Anti-Netrin G2 Rabbit Monoclonal Antibody [clone: EPR4570(2)]

Fornecedor: Abcam
AB248632-100 AB248632-1
ABCAAB248632-100EA 1020 EUR
ABCAAB248632-100 ABCAAB248632-1
Anti-Netrin G2 Rabbit Monoclonal Antibody [clone: EPR4570(2)]
Anticorpos
Rabbit monoclonal [EPR4570(2)] to Netrin G2 - BSA and Azide free.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Netrin G2
Clonality: Monoclonal
Clone: EPR4570(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR