Print…

Anti-C9 Rabbit Monoclonal Antibody [clone: EPR11232-82]

Fornecedor: Abcam
AB173302-100 AB173302-1ML
ABCAAB173302-100EA 619 EUR
ABCAAB173302-100 ABCAAB173302-1ML
Anti-C9 Rabbit Monoclonal Antibody [clone: EPR11232-82]
Anticorpos
Rabbit monoclonal [EPR11232-82] to C9.

Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: C9
Clonality: Monoclonal
Clone: EPR11232-82
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR