Print…
Anti-Cubilin Rabbit Monoclonal Antibody [clone: EPR16259-27]
Fornecedor: Abcam
AB191073-100 AB191073-1ML
ABCAAB191073-100EA 682 EUR
ABCAAB191073-100 ABCAAB191073-1ML
Anti-Cubilin Rabbit Monoclonal Antibody [clone: EPR16259-27]
Anticorpos
Rabbit monoclonal [EPR16259-27] to Cubilin - C-terminal.
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Cubilin
Clonality: Monoclonal
Clone: EPR16259-27
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human