Print…

Anti-Iba1 Goat Chimera Monoclonal Antibody [clone: EPR16588]

Fornecedor: Abcam
AB289874-100 AB289874-20UL AB289874-1ML
ABCAAB289874-100EA 475 EUR
ABCAAB289874-100 ABCAAB289874-20UL ABCAAB289874-1ML
Anti-Iba1 Goat Chimera Monoclonal Antibody [clone: EPR16588]
Anticorpos
Goat Chimera monoclonal [EPR16588] to Iba1 - Chimeric.

Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Iba1
Clonality: Monoclonal
Clone: EPR16588
Conjugation:
Epitope:
Host: Goat Chimera
Isotype: IgG
Reactivity: Human; Mouse; Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR