Print…
Anti-Lumican Rabbit Monoclonal Antibody [clone: EPR24310-47]
Fornecedor: Abcam
AB278343-100 AB278343-1
ABCAAB278343-100EA 869 EUR
ABCAAB278343-100 ABCAAB278343-1
Anti-Lumican Rabbit Monoclonal Antibody [clone: EPR24310-47]
Anticorpos
Rabbit monoclonal [EPR24310-47] to Lumican - BSA and Azide free (Detector).
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Lumican
Clonality: Monoclonal
Clone: EPR24310-47
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human