Print…

Anti-Olfactory Marker Protein/OMP Rabbit Monoclonal Antibody [clone: EPR19157]

Fornecedor: Abcam
AB250713-1 AB250713-100
ABCAAB250713-1EA 9160 EUR
ABCAAB250713-1 ABCAAB250713-100
Anti-Olfactory Marker Protein/OMP Rabbit Monoclonal Antibody [clone: EPR19157]
Anticorpos
Rabbit monoclonal [EPR19157] to Olfactory Marker Protein/OMP - BSA and Azide free.

Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Olfactory Marker Protein/OMP
Clonality: Monoclonal
Clone: EPR19157
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR