Print…
Anti-MPTX+MPTX2 Rabbit Monoclonal Antibody [clone: EPR20920-19]
Fornecedor: Abcam
AB238123-100 AB238123-1ML
ABCAAB238123-100EA 788 EUR
ABCAAB238123-100 ABCAAB238123-1ML
Anti-MPTX+MPTX2 Rabbit Monoclonal Antibody [clone: EPR20920-19]
Anticorpos
Rabbit monoclonal [EPR20920-19] to MPTX+MPTX2.
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: MPTX+MPTX2
Clonality: Monoclonal
Clone: EPR20920-19
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse