Print…

Anti-Aldolase+Aldolase C Rabbit Monoclonal Antibody [clone: EPR19355]

Fornecedor: Abcam
AB251296-100 AB251296-1
ABCAAB251296-100EA 1020 EUR
ABCAAB251296-100 ABCAAB251296-1
Anti-Aldolase+Aldolase C Rabbit Monoclonal Antibody [clone: EPR19355]
Anticorpos
Rabbit monoclonal [EPR19355] to Aldolase+Aldolase C - BSA and Azide free.

Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Aldolase+Aldolase C
Clonality: Monoclonal
Clone: EPR19355
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR