Print…

Anti-Ki67 Rabbit Polyclonal Antibody

Fornecedor: Abcam
AB230460-1 AB230460-100 AB230460-500
ABCAAB230460-1EA 1040 EUR
ABCAAB230460-1 ABCAAB230460-100 ABCAAB230460-500
Anti-Ki67 Rabbit Polyclonal Antibody
Anticorpos
Rabbit polyclonal to Ki67 - C-terminal.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Ki67
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR