Print…

2-Benzoylacetanilide ≥98.0% (por base de azoto total)

Fornecedor: TCI

B0904-25G B0904-5G
TCIAB0904-25GEA 101 EUR
TCIAB0904-25G TCIAB0904-5G
2-Benzoylacetanilide ≥98.0% (por base de azoto total)
2-Benzoylacetanilide
Fórmula: C₁₅H₁₃NO₂
Número MDL : MFCD00003084
Número CAS: 959-66-0
EINECS: 213-503-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdcc458326131b8fcc0ca5e47a63e247ddff9134&vid=fdcc458326131b8fcc0ca5e47a63e247ddff9134&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 105.0 to 108.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR