Print…
Process development columns for IEC, ToyoScreen®
Fornecedor: Tosoh Bioscience
TOYO0021952EA 285 EUR
TOYO0021952 TOYO0021951 TOYO0021367 TOYO0021366 TOYO0021860 TOYO0021871 TOYO0021365 TOYO0021364 TOYO0021870 TOYO0021363 TOYO0021859 TOYO0021869 TOYO0021362 TOYO0021395 TOYO0021361 TOYO0021394 TOYO0021393 TOYO0021371 TOYO0021360 TOYO0021370 TOYO0021392
Process development columns for IEC, ToyoScreen®
ToyoScreen® process development columns are easy-to-use, prepacked columns in cartridge format containing TOYOPEARL® resins.
  • Low cost, effi cient alternative to self packing
  • Easy connections with ÄKTA®, FPLC and HPLC
Beyond initial resin screening, ToyoScreen® columns can also be used to quickly establish optimum chromatographic conditions or to purify small amounts of the target molecule for further R&D experiments.