Print…
Upright refrigerators, Comfort
Fornecedor: Liebherr
K4220 K3620 KP3620
471-1130EA 0 EUR
471-1130 471-1129 471-1070
Upright refrigerators, Comfort
Frigoríficos
Upright general use refrigerators.
  • The MagicEye control with its visual temperature display keeps the temperature at exactly the selected level
  • The high performance forced air system ensures rapid cooling of food and an even temperature throughout the entire internal space
  • The LED ceiling light creates a high-class lighting ambience within the refrigerator
  • GlassLine storage shelves
  • Ergonomic slimline handle
  • Height-adjustable feet at the front, transport castors and transport handles at the back
  • Reversible doors