Print…
Swabs in tubes
Fornecedor: Greiner Bio-One
421161_1000 420161_1300 421008 425010_1000
391-3795EA 343 EUR
391-3795 GREI420161_1300 GREI421008 GREI425010_1000
Swabs in tubes
Zaragatoas Zaragatoas em tubos
These swab tubes are suitable for taking non-human bacteriological, serological or cytological samples.
  • Sterilised by irradiation
  • Tubes are made of polystyrene
  • Swabs are made of cotton
Suitable for hygienic controls in food industry as well as in environmental sampling and veterinary research.
Atenção: * O (s) produto (s) nesta página não se enquadra no âmbito da Diretiva da UE 2019/904