Print…

Dimethyl cyclohexane-1,4-dicarboxylate (mixture of cis and trans isomers) mistura cis- e trans- ≥98.0% (por GC)

Fornecedor: TCI
Atenção

C1154-25G C1154-500G
TCIAC1154-25GEA 16 EUR
TCIAC1154-25G TCIAC1154-500G
Dimethyl cyclohexane-1,4-dicarboxylate (mixture of cis and trans isomers) mistura cis- e trans- ≥98.0% (por GC)
Dimethyl cyclohexane-1,4-dicarboxylate (mixture of cis and trans isomers)
Fórmula: C₁₀H₁₆O₄
Número MDL : MFCD00001460
Número CAS: 94-60-0
EINECS: 202-347-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66faa299118ec0fb3ef07bf105f68dc4e801c440&vid=23580b74dcd1e3274109f8c5ace03285965e6d07&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR