Print…
Chemical resistant gloves, AlphaTec® 87-650 / 87-665
Fornecedor: Ansell
87-650/7.5-8 87-665/9.5-10 87-650/8.5-9 87-650/9.5-10 87-665/6.5-7 87-665/8.5-9 87-665/7.5-8 87-650/6.5-7
112-6442EA 0 EUR
112-6442 ANSE87-665/9.5-10 112-6443 112-6444 ANSE87-665/6.5-7 ANSE87-665/8.5-9 ANSE87-665/7.5-8 112-6441
Chemical resistant gloves, AlphaTec® 87-650 / 87-665
Luvas
These natural rubber gloves deliver excellent sensitivity combined with superior tensile strength.
  • Pure cotton flocklining is soft, non irritant and superbly comfortable
  • Specially treated to reduce the risk of allergic reactions
  • Cotton flock lining for more comfort
Ideal for light assembly tasks, oil and petrol industries, cleaning tasks, chemical handlings and chemical refining.
Certificações: EN ISO 374-1:2016 Type B (KLP), EN ISO 374-5:2016, EN 388:2016 (1010A), EN 421; Cat. III; AQL 0,65