Print…
HPLC columns, Accucore™ XL C18 and XL C8
Fornecedor: Thermo Fisher Scientific
74104-012101 74104-013001 74104-014001 74104-052130 74104-102130 74104-103030 74104-104630 74104-153030 74104-154630 74104-253030 74104-254630 74204-012101 74204-013001 74204-014001 74204-053030 74204-104630 74204-153030 74204-154630 74204-254630
554-2263EA 556 EUR
554-2263 554-2264 554-2265 554-2239 554-2240 554-2244 554-2248 554-2245 554-2249 554-2246 554-2250 554-2266 554-2267 554-2268 554-2254 554-2260 554-2256 554-2261 554-2262
HPLC columns, Accucore™ XL C18 and XL C8
Cromatografia, Colunas
Based on Core Enhanced Technology, using 4 µm solid core particles, these columns allow users of conventional HPLC methods to enjoy performance far beyond that of columns packed with 5, 4 or even 3 μm fully porous particles. Very high separation efficiencies, using standard HPLC instruments and conditions, provide increased peak resolution and lower limits of detection. An ultra-stable packed bed results in exceptionally robust columns that demonstrate excellent retention and response reproducibility.
  • Economical and easy to use
  • Compatible with Uniguard™ direct-connection and stand-alone holders
  • Recommended for analytes with moderate hydrophobicity
  • Same System, Same Method, Better Results
Accucore™ XL C18: Optimum retention of non polar compounds. Hydrophobic interaction mechanism. Separates a broad range of analytes.
Accucore™ XL C8: Similar selectivity to C18 with lower retention. Recommended for analytes with moderate hydrophobicity.