Print…

(2S,3S)-(+)-2,3-O-Isopropylidene-L-threitol ≥97.0% (por GC)

Fornecedor: TCI

Sinônimos: (+)-2,3-O-Isopropylidene-L-threitol

I0376-1G I0376-5G
TCIAI0376-1GEA 43 EUR
TCIAI0376-1G TCIAI0376-5G
(2S,3S)-(+)-2,3-O-Isopropylidene-L-threitol ≥97.0% (por GC)
(2S,3S)-(+)-2,3-O-Isopropylidene-L-threitol
Fórmula: C₇H₁₄O₄
Número MDL : MFCD00063761
Número CAS: 50622-09-8
EINECS: 256-658-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a53c3877246b84cc0c245623d1b3a2e478519155&vid=a53c3877246b84cc0c245623d1b3a2e478519155&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Melting point 42.0 to 49.0 deg-C
Specific rotation [a]20/D +3.0 to +5.0 deg (C=5, Acetone)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR