Print…

Bistrifluoroacetamide ≥98.0% (por base de azoto total)

Fornecedor: TCI
Atenção

B0986-25G B0986-5G
TCIAB0986-25GEA 317 EUR
TCIAB0986-25G TCIAB0986-5G
Bistrifluoroacetamide ≥98.0% (por base de azoto total)
Bistrifluoroacetamide
Fórmula: C₄HF₆NO₂
Ponto de ebulição: 120°C
Número MDL : MFCD00013564
Número CAS: 407-24-9
EINECS: 206-981-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbd217a985e61076cb66fbf4b78fcbc93977800d&vid=bbd217a985e61076cb66fbf4b78fcbc93977800d&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 84.0 to 89.0 deg-C
Solubility in hot Toluene almost transparency
Effect test of GC derivalization Effective as derivatization agent N-Methylaniline


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR