Print…

Benzyldimethyldodecylammonium bromide ≥97.0% (por análise titulométrica)

Fornecedor: TCI
Perigo

Sinônimos: Benzyldodecyldimethylammonium bromide, Benzalkonium bromide, N-Benzyl-N,N-dimethyldodecan-1-aminium bromide

B3748-25G B3748-5G
TCIAB3748-25GEA 345 EUR
TCIAB3748-25G TCIAB3748-5G
Benzyldimethyldodecylammonium bromide ≥97.0% (por análise titulométrica)
Benzyldimethyldodecylammonium bromide
Fórmula: C₂₁H₃₈BrN
Peso Molecular: 384.44 g/mol
Número MDL : MFCD00011768
Número CAS: 7281-04-1
EINECS: 230-698-4
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5469cf6d4ac0afb42604e407fdb9b584435b312&vid=c5469cf6d4ac0afb42604e407fdb9b584435b312&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (TPBNa Method) min. 97.0 %
Melting point 50.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR