Print…

Ethyl oxamate ≥98.0% (por base de azoto total)

Fornecedor: TCI

Sinônimos: Oxamic Acid Ethyl Ester, Ethyl 2-amino-2-oxoacetate

O0085-25G O0085-250G
TCIAO0085-25GEA 16 EUR
TCIAO0085-25G TCIAO0085-250G
Ethyl oxamate ≥98.0% (por base de azoto total)
Ethyl oxamate
Fórmula: C₄H₇NO₃
Número MDL : MFCD00008005
Número CAS: 617-36-7
EINECS: 210-512-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a21e6e4c2197c53b5b9169dad45ccabc6d6722d&vid=4a21e6e4c2197c53b5b9169dad45ccabc6d6722d&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 113.0 to 117.0 deg-C
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR