2,2-Dimethylbutane ≥98.0% (por GC)
Fornecedor: TCI
D0689-25ML
D0689-100ML
TCIAD0689-25MLEA
49
EUR
TCIAD0689-25ML
TCIAD0689-100ML
2,2-Dimethylbutane ≥98.0% (por GC)
2,2-Dimethylbutane
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |