Print…

4-Chlorobenzhydrol ≥98.0% (por GC)

Fornecedor: TCI

Sinônimos: 4-Chlorophenyl phenyl carbinol, 4-Chlorobenzhydryl alcohol, 4-Chloro-α-phenylbenzyl alcohol, p-Chlorobenzhydrol, 4-Chlorodiphenylmethanol

C0989-25G C0989-500G
TCIAC0989-25GEA 30 EUR
TCIAC0989-25G TCIAC0989-500G
4-Chlorobenzhydrol ≥98.0% (por GC)
4-Chlorobenzhydrol
Fórmula: C₁₃H₁₁ClO
Peso Molecular: 218.68 g/mol
Número MDL : MFCD00004491
Número CAS: 119-56-2
EINECS: 204-333-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72d0891eb91f54b6c1d29dcbe88c9c73a10e0ff6&vid=72d0891eb91f54b6c1d29dcbe88c9c73a10e0ff6&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 63.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR