Print…
Chemical resistant gloves, North Silver Shield® / 4H®
Fornecedor: NEW PIG
WPL262-XL WPL262-L WPL262-M
NEWPWPL262-XLEA 15.5 EUR
NEWPWPL262-XL NEWPWPL262-L NEWPWPL262-M
Chemical resistant gloves, North Silver Shield® / 4H®
Luvas
These lightweight gloves made of Norfoil® laminate provide lightweight protection from corrosive chemicals. They resist permeation and breakthrough of even the most dangerous liquids like aromatics, ketones, alcohols, esters and chlorinated and aliphatic solvents.
  • Ambidextrous design makes these ideal for single glove use - one glove can be discarded if necessary without wasting a pair
  • Light-weight glove can be used as a secondary inner glove, allowing worker maximum protection in heavy duty jobs
  • Wrist is well protected due to longer 400 mm length
  • Does not contain latex or chemical accelerators minimising the chance of allergic reactions
Typical industries and applications include Chemical, Industrial Cleaning, Medical and Pharmaceutical, Laboratories, and emergency applications where it is not clear with what chemical the user may get in contact with.
Weight: 23 g