Sulphur trioxide triethylamine complex, Sigma-Aldrich®
Fornecedor: Merck
84739-10G
84739-50G
SIAL84739-10GEA
127
EUR
SIAL84739-10G
SIAL84739-50G
Sulphur trioxide triethylamine complex, Sigma-Aldrich®
Sulphur trioxide triethylamine complex
Fórmula:
C₆H₁₅NO₃S |
Número CAS:
761-01-3 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...