Print…

Ethyl-N-hydroxyacetimidate, Sigma-Aldrich®

Fornecedor: Merck

Sinônimos: Ethyl acetohydroximate

Nível de qualidade M-Clarity™ = MQ100

208922-10G 208922-50G
SIAL208922-10GEA 147 EUR
SIAL208922-10G SIAL208922-50G
Ethyl-N-hydroxyacetimidate, Sigma-Aldrich®
Ethyl-N-hydroxyacetimidate
Fórmula: C₄H₉NO₂
Número MDL : MFCD00002114
Número CAS: 10576-12-2
EINECS: 234-165-7

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR