Print…

L(-)-Malic acid monosodium salt, Sigma-Aldrich®

Fornecedor: Merck

M1125-100G M1125-25G M1125-5G
SIALM1125-100GEA 566 EUR
SIALM1125-100G SIALM1125-25G SIALM1125-5G
L(-)-Malic acid monosodium salt, Sigma-Aldrich®
L(-)-Malic acid monosodium salt
Fórmula: C₄H₅NaO₅
Número CAS: 68303-40-2

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR