Print…

Propionylthiocholine iodide, Sigma-Aldrich®

Fornecedor: Merck

P2880-25G P2880-5G
SIALP2880-25GEA 752 EUR
SIALP2880-25G SIALP2880-5G
Propionylthiocholine iodide, Sigma-Aldrich®
Propionylthiocholine iodide
Fórmula: C₈H₁₈INOS
Número CAS: 1866-73-5
EINECS: 217-482-5

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR