Print…
Raschig rings
Fornecedor: GLASWARENFABRIK KARL HECHT
41400010 41400005 41400006 41400008 41400003 41400004
HECH41400010EA 41.2 EUR
HECH41400010 459-0157 459-0158 459-0159 HECH41400003 HECH41400004
Raschig rings
Recheios de Colunas de Destilação
Soda lime glass.
  • For packing columns
  • Good chemical resistance
  • Good thermal resistance