Print…

(1-Hexadecyl)tributylphosphonium bromide, Sigma-Aldrich®

Fornecedor: Merck

Sinônimos: Hexadecyltri-n-butylphosphonium bromide, Tributylhexadecylphosphonium bromide, Tributyl(hexadecyl)phosphonium bromide, (n-Hexadecyl)tri-n-butylphosphonium bromide

Nível de qualidade M-Clarity™ = MQ100

276200-100G 276200-25G 276200-5G
SIAL276200-100GEA 555 EUR
SIAL276200-100G SIAL276200-25G SIAL276200-5G
(1-Hexadecyl)tributylphosphonium bromide, Sigma-Aldrich®
(1-Hexadecyl)tributylphosphonium bromide
Fórmula: C₂₈H₆₀BrP
Peso Molecular: 507.65 g/mol
Número MDL : MFCD00011775
Número CAS: 14937-45-2
EINECS: 239-014-9

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR