Print…

(3-Chlorophenyl)acetic acid methyl ester ≥98.0% (por GC)

Fornecedor: TCI

Sinônimos: Methyl (3-chlorophenyl)acetate, Methyl 3-chlorophenylacetate, Methyl (3-Chlorophenyl)acetate

M2142-25G M2142-5G
TCIAM2142-25GEA 115 EUR
TCIAM2142-25G TCIAM2142-5G
(3-Chlorophenyl)acetic acid methyl ester ≥98.0% (por GC)
(3-Chlorophenyl)acetic acid methyl ester
Fórmula: (C₆H₄Cl)CH₂COOCH₃
Número MDL : MFCD00800679
Número CAS: 53088-68-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1061deb0cfbf54bee48303ee67049c057f3ba2f2&vid=1061deb0cfbf54bee48303ee67049c057f3ba2f2&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR