Print…
Rubber conical gaskets Guko, DURAN®
Fornecedor: DWK Life Sciences
292020001 292022705 292022302 292023204 292023607 292023907 292024303 292021203 292021709
511-0018EA 53.7 EUR
511-0018 511-0027 511-0026 511-0028 511-0029 SCOT292023907 SCOT292024303 511-0019 511-0025
Rubber conical gaskets Guko, DURAN®
Suportes Support Sleeves
EPDM rubber.
Conical gaskets to place between filter funnel and filter flask.