Print…
Density hydrometers for mineral oil
Fornecedor: Alla France
3400FC001/60QP 3400FC009/60QP 3400FC019/60QP 3400FC029/60QP 3400FC039/60QP 3400FC049/60QP 3400FC059/60QP 3400FC069/60QP 3400FC079/60QP 3400FC089/60QP 3410FC000/60QP 3410FC005/60QP 3410FC010/60QP 3410FC015/60QP 3410FC020/60QP 3410FC025/60QP 3410FC030/60QP 3410FC035/60QP 3410FC040/60QP 3410FC045/60QP 3410FC050/60QP 3410FC055/60QP 3410FC060/60QP 3410FC065/60QP 3410FC070/60QP 3410FC075/60QP 3410FC080/60QP 3410FC085/60QP 3410FC090/60QP 3410FC095/60QP
614-0261EA 48.6 EUR
614-0261 614-0262 614-0263 614-0264 614-0265 614-0266 614-0267 614-0268 614-0269 614-0270 614-0271 614-0272 614-0273 614-0274 614-0275 614-0276 614-0277 614-0278 614-0279 614-0280 614-0281 614-0282 614-0283 614-0284 614-0285 614-0286 614-0287 614-0288 614-0289 614-0290
Density hydrometers for mineral oil
Hidrómetros
A2LA traceable hydrometers for mineral oil graduated in API.
  • Calibration temperature of 15 or 56 °C
  • Reading below meniscus
  • Low surface tension between 16 and 35 mN/m